UniVer® 3 Hardness Reagent Powder Pillows, Hach
Supplier: Hach
96299 962-99
89230-900PK 29.56 USD
89230-900 CA962-99
UniVer® 3 Hardness Reagent Powder Pillows, Hach
Environmental Test Kits
Combined indicator/buffer for total hardness determination by titration
For use with total hardness test kits.
Ordering information: Pack of 100 Powder Pillows.