Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
sensION Platinum Series Combination pH Electrode, 5-pin connector, Hach
Supplier: Hach
5191000
97021-592EA 648.6 USD InStock
97021-592
sensION Platinum Series Combination pH Electrode, 5-pin connector, Hach
Electrodes pH Electrodes
Hach Platinum Series pH probe is a standard combination pH probe with a replaceable gel reference and built-in temperature sensor.
  • Accurate
  • Easy to Use
  • Rugged and Durable
  • Flexible
Probe comes with two electrolyte cartridges and has a fixed cable with 5-pin circular connector compatible with Hach sensION meters.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty