FerroVer® Iron Reagent Powder Pillows, Hach
Supplier: Hach
21057-69 2105769
97026-696PK 32.86 USD InStock
CA21057-69 97026-696
FerroVer® Iron Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of Iron by the 1, 10 phenanthroline method
USEPA approved method for reporting wastewater analysis using Hach Method 8008.
Ordering information: Pack of 100 powder pillows.