DPD Total Chlorine Reagent Powder Pillows, Hach
Supplier: Hach
14064-28 1406428 2105603 2105603
97026-220PK 318.87 USD
CA14064-28 97026-220 CA11024-974 10000-684
DPD Total Chlorine Reagent Powder Pillows, Hach
Environmental Test Kits
For total chlorine determination by the DPD method.
  • Boric acid free
USEPA accepted for reporting drinking and wastewater analysis using Hach Method 8167.
Ordering information: Pack of 300 and 1000 powder pillows.