DR 6000EDU™ UV-VIS Spectrophotometer, Hach
LPV4419910002
89409-698EA 10983.87 USD InStock
89409-698
DR 6000EDU™ UV-VIS Spectrophotometer, Hach
Spectrophotometers and Spectrometers UV/Visible Spectrophotometers
The simple, easy-to-operate DR 6000EDU allows you to focus your attention on the research at hand and on your students.
  • With UV and Visible Spectrum capabilities, preprogrammed and configurable methods,
  • fast wavelength scanning, and integrated quality assurance software, the DR 6000EDU ensures you are ready to handle all your analytical testing needs.
Optimized for education and research settings, the DR 6000EDU offers an intuitive interface that is both easy-to-teach and easy-to use, the ability to download data and analyze it on a separate computer, and built-in quality assurance so you and your students can be confident in your results.
Certifications: cTUVus. IP20 with closed lid.
Ordering information: One year warranty.