Potassium iodide, powder pillow

Supplier: Hach

Synonyms: Dipotassium diiodideKali iodide

107799 1077-99
97026-720PK 36.88 USD
97026-720 CA1077-99
Potassium iodide, powder pillow
Potassium iodide

Reagent for several iodometric titration methods.


Ordering information: Pack of 100 powder pillows.

Formula: Kl
MW: 166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0

Order Now

Specification Test Results

Method 10025
Method Number 2 10026
Method Number 3 8161
Method Number 4 8209
Number of tests 100
Platform Powder Pillows
Quantity 100 pk
Range Various

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR