Ammonia Cyanurate Reagent Powder Pillows, Hach
Supplier: Hach
2653199 26531-99
89231-156PK 67.85 USD
89231-156 CA26531-99
Ammonia Cyanurate Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of Ammonia Nitrogen by the Salicylate method
Range: 0.01 - 0.50mg/L NH3-N.
Hach Method 8155. 
Ordering information: Pack of 100 Powder Pillows.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.