ACE® C4 Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP – L26, Mac Mod
ACE1130501 ACE1232502 ACE1230501 ACE1232504 ACE1131202 ACE1131504 ACE1130504 ACE1131001 ACE1230201 ACE1230503 ACE1130201 ACE1231204 ACE1231246 ACE1131004 ACE1131501 ACE1230202 ACE1233502 ACE1230303 ACE1131002 ACE1233503 ACE1233504 ACE1131003 ACE1232546 ACE1231001 ACE1130246 ACE1230302 ACE1230546 ACE1230346 ACE1231504 ACE1137546 ACE1130346 ACE1131201 ACE1133503 ACE1231002 ACE1237502 ACE1237546 ACE1237501 ACE1130302 ACE1231546 ACE1131246 ACE1130303 ACE1130546 ACE1130202 ACE1130301 ACE1233501 ACE1133501 ACE1131046 ACE1130502 ACE1137502 ACE1137504 ACE1232501 ACE1231503 ACE1233546 ACE1230246 ACE1231004 ACE1231502 ACE1131204 ACE1131502 ACE1231501 ACE1131203 ACE1231046 ACE1133504 ACE1230301 ACE1237504 ACE1231202 ACE1237503 ACE1130503 ACE1230502 ACE1232503 ACE1133546 ACE1137503 ACE1231003 ACE1130203 ACE1131546 ACE1230504 ACE1231203 ACE1230203 ACE1133502 ACE1137501 ACE1131503 ACE1231201
101665-698EA 870.75 USD
101665-698 101665-816 101665-848 101665-824 101665-654 101665-668 101665-704 101665-642 101665-804 101665-856 101665-672 101665-780 101665-782 101665-648 101665-662 101665-806 101665-840 101665-834 101665-644 101665-842 101665-844 101665-646 101665-826 101665-752 101665-678 101665-832 101665-862 101665-836 101665-798 101665-716 101665-686 101665-652 101665-692 101665-756 101665-870 101665-878 101665-866 101665-682 101665-800 101665-660 101665-684 101665-706 101665-674 101665-680 101665-838 101665-688 101665-650 101665-700 101665-710 101665-714 101665-812 101665-794 101665-846 101665-810 101665-764 101665-790 101665-658 101665-664 101665-786 101665-656 101665-766 101665-694 101665-830 101665-876 101665-774 101665-874 101665-702 101665-852 101665-820 101665-696 101665-712 101665-760 101665-676 101665-670 101665-860 101665-778 101665-808 101665-690 101665-708 101665-666 101665-770
ACE® C4 Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP – L26, Mac Mod
Chromatography Columns
ACE® C4 uses a proprietary bonding and end-capping process to produce a very rugged and robust butyl (C4) phase
The ultra-inert characteristics of the ACE® Columns make them the ideal choice for separating polar, moderately polar, and non-polar analytes including uncharged acids and bases, strong acids or bases in un-ionized form or in ionized form using an ion-pair modifier. ACE® columns consistently produce measurably better peak shape and column efficiency when separating troublesome compounds.
ACE® C4 is available in 3 micron and 5 micron particles sizes. ACE® C4 has a surface area of 300 m2/g and a carbon load of 5.5%. ACE® C4 has a recommended pH range of 2-8. Working outside these ranges will reduce column lifetime. ACE® C4 is also available for UHPLC applications under the brand name ACE® EXCEL C4 2 micron UHPLC columns.
ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is a silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic separations.
Ordering information: ACE® C4 columns have two options for pre-column guard systems. One is a stand-alone guard holder which needs the guard cartridge pack (See the Accessories tab) that matches the analytical column ID size that is chosen and needs a column coupler. Most customers use the integral guard system. Guard cartridge 5-packs are sold separately. (See the Accessories tab.)