ACE® Phenyl Ultra-Inert 3 and 5 µm HPLC Columns, USP - L11, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
ACE1251201 ACE1151002 ACE1252546 ACE1251046 ACE1151201 ACE1252503 ACE1253501 ACE1151203 ACE1250303 ACE1252502 ACE1150201 ACE1251001 ACE1257502 ACE1253546 ACE1251502 ACE1252501 ACE1250546 ACE1251501 ACE1153546 ACE1151501 ACE1253503 ACE1251546 ACE1150346 ACE1151046 ACE1250202 ACE1150504 ACE1251004 ACE1251503 ACE1250302 ACE1250501 ACE1150246 ACE1150503 ACE1151502 ACE1150202 ACE1151004 ACE1250504 ACE1151504 ACE1157503 ACE1250346 ACE1150546 ACE1151204 ACE1157502 ACE1251202 ACE1157504 ACE1250503 ACE1151546 ACE1257504 ACE1251003 ACE1150302 ACE1250203 ACE1252504 ACE1250201 ACE1257546 ACE1251002 ACE1251246 ACE1250301 ACE1151003 ACE1251504 ACE1251204 ACE1250502 ACE1257501 ACE1150502 ACE1151001 ACE1153502 ACE1250246 ACE1153501 ACE1150301 ACE1151503 ACE1150501 ACE1151246 ACE1150303 ACE1153503 ACE1150203 ACE1253504 ACE1157546 ACE1251203 ACE1151202 ACE1153504 ACE1253502 ACE1257503 ACE1157501
101667-856EA 1004.71 USD InStock
101667-856 101667-730 101667-912 101667-852 101667-738 101667-906 101667-924 101667-742 101667-920 101667-902 101667-758 101667-838 101667-956 101667-932 101667-876 101667-898 101667-948 101667-872 101667-782 101667-748 101667-928 101667-886 101667-772 101667-736 101667-892 101667-790 101667-850 101667-880 101667-918 101667-934 101667-764 101667-788 101667-750 101667-760 101667-734 101667-946 101667-754 101667-798 101667-922 101667-792 101667-744 101667-796 101667-860 101667-800 101667-942 101667-756 101667-962 101667-846 101667-768 101667-894 101667-910 101667-890 101667-964 101667-842 101667-868 101667-916 101667-732 101667-884 101667-866 101667-938 101667-952 101667-786 101667-728 101667-776 101667-896 101667-774 101667-766 101667-752 101667-784 101667-746 101667-770 101667-778 101667-762 101667-930 101667-802 101667-864 101667-740 101667-780 101667-926 101667-960 101667-794
ACE® Phenyl Ultra-Inert 3 and 5 µm HPLC Columns, USP - L11, Mac Mod
Chromatography Columns
ACE® Phenyl uses a proprietary bonding and end-capping process to produce a very rugged and robust alkylphenyl phase
ACE® Phenyl has a hydrophobicity that is between C4 and C8 phases, with increased polar selectivity. ACE® Phenyl has improved performance, stability and reproducibility compared to conventional phenyl phases. ACE® Phenyl is compatible with highly aqueous mobile phases.
ACE® Phenyl is available in 3 micron and 5 micron particles sizes. ACE® Phenyl has a surface area of 300 m2/g and a carbon load of 9.5%. ACE® Phenyl has a recommended pH range of 2-8. Working outside these ranges will reduce column lifetime. ACE® Phenyl is also available for UHPLC applications under the brand name ACE® EXCEL Phenyl 2 micron UHPLC columns.
ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic separations
Ordering information: ACE® Phenyl columns have two options for pre-column guard systems. One is a stand-alone guard holder which needs the guard cartridge pack (See the Accessory tab on this page) that matches the analytical column ID size that is chosen and needs a column coupler. Most customers use the integral guard system.