Would you like to log in again?
ACE® CN Ultra-Inert, 5 and 10 µm, Semi-Preparative and Preparative HPLC Columns, USP - L10, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
ACE1241020 ACE1241508 ACE1241008 ACE1242510 ACE1347520 ACE1347508 ACE1347546 ACE1241010 ACE1247520 ACE1240520 ACE1342530 ACE1340346 ACE1340508 ACE1340546 ACE1242520 ACE1341008 ACE1241210 ACE1241030 ACE1341020 ACE1341010 ACE1341210 ACE1341246 ACE1341030 ACE1341546 ACE1242530 ACE1343546 ACE1341510 ACE1240508 ACE1340246 ACE1342508 ACE1241520 ACE1342546 ACE1241530 ACE1341520 ACE1242508 ACE1347510 ACE1341046 ACE1247510 ACE1340520 ACE1341208 ACE1341220 ACE1341530 ACE1241208 ACE1241510 ACE1340510 ACE1240530 ACE1341508 ACE1247508 ACE1240510 ACE1342520 ACE1342510 ACE1241220
101667-142EA 3676.51 USD InStock
101667-142 101667-186 101667-152 101667-198 101667-012 101667-016 101667-014 101667-138 101667-256 101667-238 101666-990 101666-996 101667-008 101667-006 101667-202 101666-964 101667-156 101667-146 101666-958 101666-956 101666-966 101666-970 101666-960 101666-980 101667-206 101666-998 101666-974 101667-248 101666-984 101666-994 101667-176 101666-992 101667-180 101666-976 101667-212 101667-010 101666-962 101667-252 101667-002 101666-972 101666-968 101666-978 101667-168 101667-172 101667-000 101667-242 101666-982 101667-264 101667-234 101666-988 101666-986 101667-160
ACE® CN Ultra-Inert, 5 and 10 µm, Semi-Preparative and Preparative HPLC Columns, USP - L10, Mac Mod
Chromatography Columns
ACE® CN uses a proprietary bonding and end-capping process to produce a very rugged and robust cyanopropyl phase
ACE® CN semi-prep and prep HPLC Columns are suitable for use in both normal- and reversed-phase modes. Greatly improved performance, stability and reproducibility compared to conventional CN phases.
ACE® CN Semi-Prep and Prep Columns are available in 5 micron and 10 micron particles sizes. ACE® C18 Semi-Prep and Prep Columns have a surface area of 300 m2/g and a carbon load of 5.5%. ACE® CN Semi-Prep and Prep Columns have a recommended pH range of 2-8. Working outside these ranges will reduce column lifetime.
ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic semi-prep and prep separations.
Ordering information: ACE® CN Semi-Prep and Prep columns can be equipped with pre-column guard systems. The guard cartridge pack matches the analytical column ID size that is chosen (See the Accessories tab on this page).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty