ACE® C18-PFP Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP - L1, Mac Mod
ACE11107503 ACE11100246 ACE12100246 ACE12102502 ACE12100502 ACE11102546 ACE11101504 ACE11101004 ACE11101503 ACE12101503 ACE12101204 ACE11100502 ACE12100202 ACE11107502 ACE11101201 ACE11101204 ACE12100203 ACE12101003 ACE12102503 ACE13107546 ACE12100503 ACE12100301 ACE12101546 ACE12102546 ACE11101501 ACE12107546 ACE12101504 ACE12103501 ACE12101001 ACE11100202 ACE12107502 ACE12102501 ACE11100302 ACE12100546 ACE12101501 ACE12100302 ACE11103502 ACE12100504 ACE11100346 ACE12101502 ACE11103501 ACE12103502 ACE11101002 ACE11100504 ACE13101246 ACE11103504 ACE12101002 ACE11100503 ACE12101203 ACE12107501 ACE12101201 ACE11101246 ACE12101246 ACE11100301 ACE11101001 ACE11101203 ACE11100546 ACE11100203 ACE12100346 ACE12101046 ACE11101046 ACE12101202 ACE12101004 ACE11103546 ACE11100303 ACE11107504 ACE12107504 ACE12100501 ACE12107503 ACE11107501 ACE11100501 ACE11101202 ACE11101003 ACE11101502 ACE12103504 ACE11107546 ACE12100303 ACE12102504 ACE11103503 ACE12103503 ACE11101546
101665-500EA 974.94 USD InStock
101665-500 101665-478 101665-534 101665-538 101665-554 101717-216 101717-210 101717-194 101665-510 101665-524 101717-270 101665-490 101665-530 101665-498 101717-198 101717-204 101665-532 101665-516 101665-540 101717-466 101665-556 101665-544 101665-526 101665-542 101665-466 101665-566 101717-274 101717-288 101665-512 101665-474 101665-562 101665-536 101665-482 101665-558 101665-520 101665-546 101717-222 101717-252 101665-486 101665-522 101717-220 101717-290 101665-460 101717-188 101717-446 101717-226 101665-514 101665-492 101717-268 101665-560 101717-264 101717-206 101717-272 101665-480 101665-458 101717-202 101665-494 101665-476 101665-550 101665-518 101665-464 101717-266 101717-258 101717-228 101665-484 101717-230 101717-300 101665-552 101665-564 101665-496 101665-488 101717-200 101665-462 101665-468 101717-294 101665-502 101665-548 101717-280 101717-224 101717-292 101665-470
ACE® C18-PFP Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP - L1, Mac Mod
Chromatography Columns
ACE® C18-PFP uses a proprietary bonding and end-capping process to produce a very rugged and robust octadecyl-pentafluorophenyl (C18-PFP) single phase
The ACE® C18-PFP offers multiple types of analyte-stationary phase interactions (hydrophobic, pi-pi, dipole-dipole, and hydrogen bonding) to deliver unique selectivity, complementary to that of the ACE® C18-AR. The ultra-inert characteristics of the ACE® C18-PFP Columns make them the ideal choice for separating analytes that differ in hydrophobicity, electron-rich analytes with electron delocalization and electron donating groups (i.e. phenols, aromatic ehters, and amines). Recommended applications for sterioisomers, steroids, taxanes, substituted aromatics, and nucleosides. ACE® columns consistently produce measurably better peak shape and column efficiency when separating troublesome compounds. ACE® C18-PFP is also compatible with 100% Aqueous mobile phases for enhanced retention in reversed phase mode.
ACE® C18-PFP is available in 3 micron and 5 micron particles sizes. ACE® C18-PFP has a surface area of 300 m2/g and a carbon load of 14.3%. ACE® C18-PFP has a recommended pH range of 2-8; working outside these ranges will reduce column lifetime. ACE® C18-PFP is also available for UHPLC applications under the brand name ACE® EXCEL C18-PFP 2 micron UHPLC columns.
ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic separations.
Ordering information: ACE® C18-PFP columns have two options for pre-column guard systems. One is a stand-alone guard holder which needs the guard cartridge pack (See the Accessory tab on this page) that matches the analytical column ID size that is chosen and needs a column coupler. Most customers use the integral guard system.