Would you like to log in again?
Digital Logic Trainer
Supplier: GLOBAL SPECIALTIES SE
DL-010
470137-748EA 412.79 USD InStock
470137-748
Digital Logic Trainer
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Allows Independent Exploration of Logic and Systems Design
Students can go right to work with this independent trainer that has eliminated the need to discreetly wire ICs. The attractive and easy to use system draws students in to the beauty of digital logic and embedded systems design. Eleven hands-on labs are included in the introductory digital logic course, complete with lab manual authored by a university professor.Students can create more than a dozen standard logic circuits and custom circuits using standard TTL logic, and the expandable breadboard allows the system to grow as students' knowledge increases. Kit includes 100 machined pin hookup wires, a 9V wall adapter, gate elements, expandable breadboard and a rugged carrying case with commonly-referenced truth table inside the case lid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty