Would you like to log in again?
ACE® C18-AR Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP - L1, Mac Mod
Supplier: MAC-MOD ANALYTICAL MS
ACE1197504 ACE1291003 ACE1290501 ACE1290346 ACE1190503 ACE1292503 ACE1191046 ACE1291046 ACE1291004 ACE1297503 ACE1190246 ACE1297501 ACE1197546 ACE1190301 ACE1190502 ACE1191503 ACE1291502 ACE1291503 ACE1191004 ACE1291504 ACE1297502 ACE1290202 ACE1190202 ACE1191504 ACE1197503 ACE1191502 ACE1292501 ACE1290504 ACE1191001 ACE1290302 ACE1290503 ACE1297504 ACE1191546 ACE1192546 ACE1197501 ACE1291001 ACE1291501 ACE1190203 ACE1290546 ACE1191002 ACE1191003 ACE1297546 ACE1191501 ACE1292502 ACE1190302 ACE1190546 ACE1290502 ACE1290246 ACE1190303 ACE1290303 ACE1190346 ACE1291002 ACE1190501 ACE1292546 ACE1290301 ACE1291546 ACE1190504 ACE1290203 ACE1197502 ACE1292504
101717-246EA 974.94 USD InStock
101717-246 101665-110 101665-146 101665-144 101665-086 101665-134 101665-058 101665-112 101717-402 101665-158 101665-072 101665-154 101665-096 101665-074 101665-084 101665-104 101665-116 101665-118 101717-236 101717-408 101665-156 101665-124 101665-068 101717-240 101665-094 101665-062 101665-130 101717-396 101665-052 101665-140 101665-150 101717-420 101665-064 101717-244 101665-090 101665-106 101665-114 101665-070 101665-152 101665-054 101665-056 101665-160 101665-060 101665-132 101665-076 101665-088 101665-148 101665-128 101665-078 101665-142 101665-080 101665-108 101665-082 101665-136 101665-138 101665-120 101717-234 101665-126 101665-092 101717-414
ACE® C18-AR Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP - L1, Mac Mod
Chromatography Columns
ACE® C18-AR uses a proprietary bonding and end-capping process to produce a very rugged and robust octadecyl-phenyl (C18-AR) single phase
The ACE® C18-AR leverages the selectivity of a C18 and a phenyl phase to separate mixtures that are very difficult to separate with either single phase alone. The ultra-inert characteristics of the ACE® C18-AR Columns make them the ideal choice for separating analytes that differ in hydrophobicity, electron-poor analytes with electron delocalization and electron withdrawing groups (i.e. halogens, nitro groups, ketones, esters, and acids). Recommended applications for basic analytes, heterocycles, proton donors and acceptors, and halogenated aromatics. ACE® columns consistently produce measurably better peak shape and column efficiency when separating troublesome compounds. ACE® C18-AR is also compatible with 100% Aqueous mobile phases for enhanced retention in reversed phase mode.
ACE® C18-AR is available in 3 micron and 5 micron particles sizes. ACE® C18-AR has a surface area of 300 m2/g and a carbon load of 15.5%. ACE® C18 has a recommended pH range of 2-8; working outside these ranges will reduce column lifetime. ACE® C18-AR is also available for UHPLC applications under the brand name ACE® EXCEL C18-AR 2 micron UHPLC columns.
ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic separations.
Ordering information: ACE® C18-AR columns have two options for pre-column guard systems. One is a stand-alone guard holder which needs the guard cartridge pack (See the Accessory tab on this page) that matches the analytical column ID size that is chosen and needs a column coupler. Most customers use the integral guard system.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty