ACE® C8 Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP – L7, Mac Mod
ACE1120503 ACE1121002 ACE1120202 ACE1221002 ACE1120203 ACE1220201 ACE1121503A ACE1121546 ACE1123546 ACE1221003 ACE1221202 ACE1220303 ACE1120303 ACE1121204 ACE1227504 ACE1222504 ACE1123503 ACE1127503 ACE1121501 ACE1220504 ACE1220502 ACE1122546 ACE1220301 ACE1221203 ACE1121202 ACE1223504 ACE1121503 ACE1123504 ACE1121504 ACE1223503 ACE1221046 ACE1120546 ACE1223502 ACE1121003 ACE1227502 ACE1221001 ACE1222503 ACE1221501 ACE1127546 ACE1120501 ACE1127504 ACE1227501 ACE1221004 ACE1221502 ACE1220302 ACE1220246 ACE1121246 ACE1221503 ACE1223546 ACE1221546 ACE1120302 ACE1123502 ACE1121001 ACE1220202 ACE1227503 ACE1220546 ACE1121502 ACE1222546 ACE1120201 ACE1227546 ACE1220203 ACE1221504 ACE1127501 ACE1121004 ACE1222502 ACE1223501 ACE1123501 ACE1121046 ACE1220503 ACE1120246 ACE1121203 ACE1120504 ACE1127502 ACE1220501 ACE1222501 ACE1120502 ACE1220346 ACE1221201 ACE1120301 ACE1120346 ACE1121201 ACE1221246
101666-394EA 870.75 USD
101666-394 101666-334 101666-364 101666-448 101666-366 101666-496 101666-620 101666-360 101666-386 101666-452 101666-466 101666-526 101666-374 101666-348 101666-568 101666-516 101666-382 101666-404 101666-352 101666-552 101666-544 101666-388 101666-522 101666-470 101666-344 101666-536 101666-356 101666-384 101666-358 101666-534 101666-458 101666-398 101666-532 101666-336 101666-562 101666-444 101666-512 101666-478 101666-408 101666-390 101666-406 101666-558 101666-456 101666-482 101666-524 101666-502 101666-350 101666-486 101666-538 101666-492 101666-372 101666-380 101666-332 101666-498 101666-566 101666-554 101666-354 101666-518 101666-362 101666-570 101666-500 101666-490 101666-400 101666-338 101666-508 101666-530 101666-378 101666-340 101666-548 101666-368 101666-346 101666-396 101666-402 101666-540 101666-504 101666-392 101666-528 101666-462 101666-370 101666-376 101666-342 101666-474
ACE® C8 Ultra-Inert, 3 and 5 µm, Analytical HPLC Columns, USP – L7, Mac Mod
Chromatography Columns
ACE® C8 uses a proprietary bonding and end-capping process to produce a very rugged and robust octyl (C8) phase
The ultra-inert characteristics of the ACE® Columns make them the ideal choice for separating polar, moderately polar, and non-polar analytes including uncharged acids and bases, strong acids or bases in un-ionized form or in ionized form using an ion-pair modifier. ACE® columns consistently produce measurably better peak shape and column efficiency when separating troublesome compounds.
ACE® C8 is available in 3 micron and 5 micron particles sizes. ACE® C8 has a surface area of 300 m2/g and a carbon load of 9.0%. ACE® C8 has a recommended pH range of 2-8. Working outside these ranges will reduce column lifetime. ACE® C8 is also available for UHPLC applications under the brand name ACE® EXCEL C8 2 micron UHPLC columns.
ACE® C8 has an increased bonding density as compared to ACE® C18. ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is a silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic separations
Ordering information: ACE® C8 columns have two options for pre-column guard systems. One is a stand-alone guard holder which needs the guard cartridge pack (See the Accessories tab) that matches the analytical column ID size that is chosen and needs a column coupler. Most customers use the integral guard system. Guard cartridge 5-packs are sold separately. (See the Accessories tab.)
Part number 101666-620 is shipped in Acetonitrile.