ACE® C18 HL Ultra-Inert, 3 and 5 µm, HPLC Columns, USP - L1, Mac Mod
ACE3111202 ACE3210246 ACE3211201 ACE3113502 ACE3213546 ACE3111501 ACE3110303 ACE3111002 ACE3211003 ACE3211546 ACE3211001 ACE3210203 ACE3117503 ACE3210302 ACE3211502 ACE3211501 ACE3211246 ACE3210546 ACE3117501 ACE3111001 ACE3211503 ACE3211046 ACE3111503 ACE3210202 ACE3211202 ACE3213504 ACE3212501 ACE3117504 ACE3212504 ACE3110302 ACE3111004 ACE3212503 ACE3117502 ACE3210346 ACE3212502 ACE3111201 ACE3111003 ACE3210301 ACE3211204 ACE3217501 ACE3210502 ACE3217503 ACE3210501 ACE3217502 ACE3110504 ACE3213501 ACE3110246 ACE3211203 ACE3111502 ACE3113503 ACE3110346 ACE3113501 ACE3110502 ACE3111046 ACE3213502 ACE3217504 ACE3111504 ACE3110201 ACE3117546 ACE3211004 ACE3213503 ACE3110202 ACE3113504 ACE3211504 ACE3212546 ACE3211002 ACE3111246 ACE3110203 ACE3210201 ACE3110546 ACE3217546 ACE3210504 ACE3111203 ACE3210303 ACE3110301 ACE3210503 ACE3113546 ACE3110501 ACE3111204
101665-248EA 1138.68 USD
101665-248 101665-376 101665-336 101665-284 101665-412 101665-256 101665-278 101665-238 101665-326 101665-366 101665-318 101665-374 101665-306 101665-398 101665-356 101665-352 101665-348 101665-428 101665-302 101665-236 101665-360 101665-332 101665-260 101665-372 101665-340 101665-410 101665-378 101665-308 101665-390 101665-276 101665-242 101665-386 101665-304 101665-402 101665-382 101665-246 101665-240 101665-396 101665-346 101665-432 101665-418 101665-440 101665-414 101665-436 101665-298 101665-404 101665-272 101665-344 101665-258 101665-286 101665-280 101665-282 101665-294 101665-244 101665-406 101665-442 101665-262 101665-266 101665-310 101665-330 101665-408 101665-268 101665-288 101665-364 101665-392 101665-322 101665-254 101665-270 101665-370 101665-300 101665-444 101665-426 101665-250 101665-400 101665-274 101665-422 101665-290 101665-292 101665-252
ACE® C18 HL Ultra-Inert, 3 and 5 µm, HPLC Columns, USP - L1, Mac Mod
Chromatography Columns
ACE® C18 HL (high load) uses a proprietary bonding and end-capping process to produce a very rugged and robust octadecyl (C18) phase with high surface area
ACE® C18 HL is a high surface area, high carbon load phase, leading to increased retention and loading compared to ACE® C18. Availability of particle sizes up to 15 microns ensures easy scale-up for preparative and process scale applications.
ACE® C18 HL is available in 3, 5, 10 and 15 micron particles sizes. ACE® C18 HL has a surface area of 400 m2/g and a carbon load of 20.0%. ACE® C18 HL has a recommended pH range of 2-8 (working outside these ranges will reduce column lifetime). ACE® C18 is also available for UHPLC applications under the brand name ACE® EXCEL C18 2 micron UHPLC columns.
ACE® C18 HL is especially useful for preparative applications. ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic separations
Ordering information: ACE® C18-HL columns have two options for pre-column guard systems. One is a stand-alone guard holder which needs the guard cartridge pack (See the Accessory tab on this page) that matches the analytical column ID size that is chosen and needs a column coupler. Most customers use the integral guard system.