ACE® AQ Ultra-Inert HPLC Columns, USP - L1, 3 and 5 µm, Mac Mod
ACE1160501 ACE1261004 ACE1161002 ACE1260202 ACE1260203 ACE1261203 ACE1260502 ACE1160346 ACE1161202 ACE1167501 ACE1262502 ACE1263504 ACE1261046 ACE1261503 ACE1263502 ACE1160504 ACE1160302 ACE1160246 ACE1167502 ACE1261003 ACE1160201 ACE1260201 ACE1167546 ACE1260501 ACE1161046 ACE1167503 ACE1161201 ACE1267546 ACE1263501 ACE1261201 ACE1261502 ACE1163503 ACE1163502 ACE1267503 ACE1260504 ACE1167504 ACE1160303 ACE1260503 ACE1263503 ACE1262501 ACE1267502 ACE1161204 ACE1261002 ACE1260346 ACE1161504 ACE1161203 ACE1267501 ACE1160202 ACE1267504 ACE1163501 ACE1163504 ACE1261202 ACE1260246 ACE1263546 ACE1260303 ACE1262504 ACE1261246 ACE1161246 ACE1261204 ACE1161001 ACE1161004 ACE1160301 ACE1163546 ACE1161003 ACE1260301 ACE1261001 ACE1161501 ACE1260302 ACE1262546 ACE1261504 ACE1160203 ACE1261501
101664-088EA 1138.68 USD
101664-088 101664-286 101664-120 101664-122 101664-084 101664-282 101664-086 101664-206 101664-128 101664-202 101664-246 101664-124 101664-204 101664-248 101664-126 101664-090 101664-290 101664-092 101664-110 101664-198 101664-230 101664-274 101664-076 101664-112 101664-078 101664-232 101664-194 101664-270 101664-118 101664-238 101664-114 101664-278 101664-234 101664-116 101664-080 101664-082 101664-264 101664-266 101664-222 101664-188 101664-100 101664-260 101664-262 101664-184 101664-228 101664-106 101664-108 101664-268 101664-102 101664-226 101664-104 101664-190 101664-132 101664-176 101664-098 101664-296 101664-252 101664-134 101664-298 101664-254 101664-210 101664-094 101664-130 101664-294 101664-096 101664-218 101664-136 101664-256 101664-138 101664-214 101664-258 101664-180
ACE® AQ Ultra-Inert HPLC Columns, USP - L1, 3 and 5 µm, Mac Mod
ACE® AQ uses a proprietary bonding and end-capping process to produce a very rugged and robust aqueous campatible phase.
ACE® AQ is a unique C18 bonded phase with integral polar functionality. Resistant to retention loss (due to pore dewetting, formerly called 'phase collapse') even with 100% aqueous mobile phases.
ACE® AQ is available in 3 and 5 µm particles sizes. ACE® AQ has a surface area of 300 m²/g and a carbon load of 14%. ACE® AQ has a recommended pH range of 2 to 8. Working outside these ranges will reduce column lifetime. ACE® AQ is also available for UHPLC applications under the brand name ACE® EXCEL AQ 2 µm UHPLC columns.
ACE® columns are manufactured using ultra-pure based-deactivated silica that has extremely low silanol activity. This ultra-pure silica is efficiently bonded and exhaustively end-capped using proprietary technology. The result is silica based stationary phase that has virtually eliminated the negative effects of silanols on chromatographic separations.
Ordering information: ACE® AQ columns have two options for pre-column guard systems. One is a stand-alone guard holder which needs the guard cartridge pack (See the Accessory tab on this page) that matches the analytical column Int. Ø size that is chosen and needs a column coupler. Most customers use the integral guard system.