PerfeCTa® qPCR FastMix® II, ROX™, QuantaBio
Supplier: Quantabio
95119-05K 95119-012 95119-250 95119-05K 95119-250 95119-012
97065-998EA 2366.24 USD
97065-998 97065-996 97066-000 CA97065-998 CA97066-000 CA97065-996
PerfeCTa® qPCR FastMix® II, ROX™, QuantaBio
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
PerfeCTa FastMix II, ROX is an advanced qPCR reagent system for both fast and conventional PCR cycling protocols or instruments
A key component of PerfeCTa FastMix II, ROX is an ultra pure, processive thermostable DNA polymerase that is free of detectable E. coli DNA§. PerfeCTa FastMix II, ROX is ideal for demanding qPCR applications such as bacterial pathogen detection where residual host DNA in typical recombinant enzyme preparations can limit assay sensitivity and obscure detection of low copy samples. The enzyme in PerfeCTa FastMix II, ROX is combined with high avidity monoclonal antibodies to provide a stringent automatic hot-start that allows reaction assembly, and temporary storage, at room temperature prior to PCR amplification. Real-Time PCR Instrument Compatibility:
Applied Biosystems 5700
Applied Biosystems 7000
Applied Biosystems 7300
Applied Biosystems 7700
Applied Biosystems 7900
Applied Biosystems 7900HT
Applied Biosystems 7900 HT Fast
Applied Biosystems StepOne™
Applied Biosystems StepOnePlus™
Contents:
2X reaction buffer containing optimized concentrations of MgCl2, dNTPs (dATP, dCTP, dGTP, dTTP), hot-start DNA polymerase, ROX reference dye, AccuVue blue qPCR dye, and stabilizers.
Storage conditions:
PerfeCTa FastMix II, ROX is stable for 2 years when stored in a constant temperature freezer at –20°C. For convenience, it may be stored unfrozen at 2 to 8°C for up to 6 months. After thawing, mix thoroughly before using. Repeated freezing and thawing does not affect PCR performance.
Precautions and Disclaimer:
For research use only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
It is a versatile and robust solution that provides the ultimate sensitivity and high PCR efficiency using a variety of fluorogenic probe chemistries, including TaqMan® hydrolysis probes. PerfeCTa FastMix II is provided as a 2X concentrated ready-to-use reaction cocktail that contains all required reaction components, except primers, probe(s), and DNA template. The light blue color of the AccuVue™ tracer dye simplifies reaction assembly in white, or clear, plates and helps to minimize pipetting or mixing errors. It does not interfere with qPCR performance or affect the stability of the product.