Would you like to log in again?
PerfeCTa® Universal PCR Primer, QuantaBio
Supplier: Quantabio
95109-500 95109-500
89168-792EA 86.18 USD InStock
CA89168-792 89168-792
PerfeCTa® Universal PCR Primer, QuantaBio
Nucleic Acid Reagents PCR Reagents Primers and Probes
The PerfeCTa Universal PCR Primer has been designed and validated to work specifically with PerfeCTa MicroRNA Assays and PerfeCTa SYBR Green SuperMix using microRNA cDNA produced using the qScript MicroRNA cDNA Synthesis Kit as template (see the relevant products section below).
MicroRNAs are not polyadenylated in nature. With the qScript MicroRNA cDNA Synthesis Kit miRNAs are polyadenylated in a poly(A) polymerase reaction. qScript Reverse Transcriptase and other necessary reagents for cDNA synthesis are subsequently added to convert the poly(A) tailed miRNAs into cDNA using an oligo-dT adapter primer. The adapter primer has a unique sequence at its 5’ end which allows amplification of cDNAs in real-time qRT-PCR reactions.
Individual microRNAs are quantitated in real-time qRT-PCR amplification reactions using the PerfeCTa microRNA Assays along with the PerfeCTa Universal PCR Primer (specific to the unique sequence of the oligo-dT adapter primer) and the PerfeCTa SYBR Green SuperMix family of products. The pre-designed and validated PerfeCTa MicroRNA Assays provide maximum sensitivity and specificity in qRT-PCR amplifications
Storage conditions:
The PerfeCTa Universal PCR Primer is stable for > one year when stored at +4°C.
Precautions and Disclaimer:
For research use only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty