Would you like to log in again?
qScript™ One-Step SYBR® Green qRT-PCR Kit, Low ROX™, QuantaBio
Supplier: Quantabio
95089-200 95089-200 95089-050 95089-050
95054-954EA 559.65 USD InStock
95054-954 CA95054-954 95054-952 CA95054-952
qScript™ One-Step SYBR® Green qRT-PCR Kit, Low ROX™, QuantaBio
Nucleic Acid Reagents PCR Reagents PCR Kits
The qScript™One-Step SYBR® Green qRT-PCR Kit, Low ROX™ is a convenient and highly sensitive solution for quantitative RT-PCR of RNA templates (qRT-PCR) using SYBR® Green I dye detection and gene-specific primers on Applied Biosystems 7500 or Stratagene MX series of real-time PCR systems
Real-Time PCR Instrument Compatibility:
Applied Biosystems 7500
Applied Biosystems 7500 Fast
Stratagene Mx3000P®
Stratagene Mx3005P™
Stratagene Mx4000™
Applied Biosystems ViiA 7
Applied Biosystems QuantStudio™ 12K Flex Contents:
- qScript One-Step Reverse Transcriptase - Optimized 50X formulation of recombinant MMLV reverse transcriptase for one-step RT-PCR.
- One-Step SYBR Green Master Mix, Low ROX (2X) - 2X reaction buffer containing dNTPs, magnesium chloride, AccuStart Taq DNA polymerase, stabilizers, ROX reference dye and SYBR Green I dye
- Nuclease-free water.
Storage conditions:
Kit components are stable for one year when stored in a constant temperature freezer at 20ºC, protected from light. For convenience, the One-Step SYBR Green Master Mix, Low ROX may be stored unfrozen at +2 to +8ºC for up to 4 months.
Precautions and Disclaimer:
For research use only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
For research use only. Not intended for any animal or human therapeutic or diagnostic use. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
cDNA synthesis and PCR amplification are carried out in the same tube without opening between procedures. The system has been optimized to deliver maximum RT PCR efficiency, sensitivity, and specificity. The proprietary reaction buffer has been specifically formulated to maximize activities of both reverse transcriptase and Taq DNA polymerase while minimizing the potential for primer-dimer and other non-specific PCR artifacts. The kit is compatible with both fast and standard qPCR cycling protocols. Highly specific amplification is essential for successful qRT-PCR with SYBR® Green I technology, since this dye binds to any dsDNA generated during amplification. AccuStart™ Taq DNA polymerase contains monoclonal antibodies that bind to the polymerase and keep it inactive prior to the initial PCR denaturation step. Upon heat activation at 95ºC, the antibodies denature irreversibly, releasing fully active, unmodified Taq DNA polymerase.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty