Would you like to log in again?
Standard-Duty Dry Diaphragm Vacuum Pumps, High Capacity, Welch-Ilmvac
Supplier: WELCH VACUUM BY GARDNER DENVER
2154B-01
89410-236EA 4036.01 USD InStock
89410-236
Standard-Duty Dry Diaphragm Vacuum Pumps, High Capacity, Welch-Ilmvac
Pumps Vacuum Pumps
Use affordable Welch-Ilmvac dry diaphragm pumps to provide oil-free vacuum for a wide variety of applications
Welch-Ilmvac diaphragm vacuum pumps are used world-wide in industry, laboratory, and education. Economical dry diaphragm pumps are quiet and ecologically superior to oil-lubricated pumps or water aspirator-generated vacuum. For applications requiring rough to medium-high vacuum (1 to 760torr). Use standard duty pump models with aqueous/non-aggressive fumes.
Select from a broad range of flow and ultimate vacuum level models to provide reliable, low-maintenance vacuum for routine and specialized needs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty