Vibration Meter and Laser Combination Tachometer, Extech
461880 461880-NIST
10171-756EA 1510.5 USD
10171-756 97050-008
Vibration Meter and Laser Combination Tachometer, Extech
Vibration Meter + Laser/Contact Tachometer that measures Acceleration, Velocity and Displacement plus RPM and Linear Surface Speed
This meter features both Contact and Non-Contact (Laser) tachometer functions and can Manual/Auto store/recall up to 1000 readings. It also Sports a large LCD with characters on display reverse direction depending on contact, photo, or vibration mode.
Certifications: CE marked.
Ordering information: Vibration sensor with 39" cable, magentic mounts, wheels (cones and flat tips), relfective tape, 4 AA batteries, software with cable, and hard carrying case included. 1 year warranty.