4-Channel Vibration Meter/Datalogger, Extech
VB500 VB500-NIST
10171-778EA 6067.81 USD
10171-778 89370-194
4-Channel Vibration Meter/Datalogger, Extech
4-Channel Vibration Meter/Datalogger that datalogs and stores readings on an SD card in Excel® format
This meter features a large backlit LCD display that simultaneously displays four channels. It also has RMS or Peak Value measurement modes, as well as Data Hold and Auto Power Off functions.
Certifications: CE marked.
Ordering information: 4 remote sensors, magnetic mounts, 8 AA batteries, SD Card, 100-240V adaptor, and durable carrying case included. 1 year warranty.