Calpain Inhibitor IV (Z-LLY-FMK), Powder

Supplier: Alfa Aesar

Synonyms: Z-Leu-Leu-Tyr-FMKZ-Leu-Leu-Tyr-Fluoromethylketonebenzyl N-[1-[[1-[[4-fluoro-1-(4-hydroxyphenyl)-3-oxobutan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]amino]-4-methyl-1-oxopentan-2-yl]carbamateZ-Leu-Leu-Tyr-fluoromethyl ketone

J65052-MCR J65052-MCR
AAJ65052-MCREA 177.22 USD InStock
CAAAJ65052-MCR AAJ65052-MCR
Calpain Inhibitor IV (Z-LLY-FMK), Powder
Calpain Inhibitor IV (Z-LLY-FMK)

A potent, cell-permeable, and irreversible inhibitor of calpain II. Also inhibits cathepsin L


  • Appearance: Yellow
  • Moisture-sensitive
  • Soluble in DMSO

Formula: C₃₀H₄₀FN₃O₆
MW: 557,7 g/mol
Storage Temperature: Freezer
MDL Number: MFCD03453605
CAS Number: 133410-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR