PTP1B inhibitor, solid

Supplier: Thermo Scientific Chemicals

J64512-LB0 J64512.LB0
AAJ64512-LB0EA 501.18 USD
CAAAJ64512-LB0 AAJ64512-LB0
PTP1B inhibitor, solid
PTP1B inhibitor

White solid


  • Light Sensitive


PTP1B inhibitor blocks the activity of the Protein Tyrosine Phosphatase 1B, which is a key factor in the negative regulation of insulin and leptin signaling pathway


Soluble in ethanol and DMSO

Formula: C₂₆H₁₉Br₂N₃O₇S₃
MW: 741.45 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09037556
CAS Number: 765317-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR