Syk inhibitor, solid

Supplier: Thermo Scientific Chemicals
Warning

J64281-LB0 J64281-MCR J64281.LB0 J64281-MCR
AAJ64281-MCREA 60.9 USD
AAJ64281-MCR AAJ64281-LB0 CAAAJ64281-MCR CAAAJ64281-LB0
Syk inhibitor, solid
Syk inhibitor

Dark orange solid


  • Light Sensitive


A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor


Soluble in DMSO

Formula: C₁₈H₁₅N₃O₃S
Storage Temperature: Refrigerator
MDL Number: MFCD07776820
CAS Number: 622387-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR