Pifithrin-α, p-Nitro hydrobromide, solid

Supplier: Thermo Scientific Chemicals

Synonyms: p-nitro-Pifithrin-α

J64277-LB0 J64277-LB0
AAJ64277-LB0EA 110.57 USD
AAJ64277-LB0 CAAAJ64277-LB0
Pifithrin-α, p-Nitro hydrobromide, solid
Pifithrin-alpha, p-Nitro hydrobromide

Pale brown solid


  • Light Sensitive
  • Hygroscopic


A cell-permeable p53 inhibitor


Soluble in DMSO or ethanol

Formula: C₁₅H₁₆BrN₃O₃S·HBr
MW: 398.30 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00398180
CAS Number: 389850-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR