MAC 1753

J64242-LB0 J64242-LB0
AAJ64242-LB0EA 193.46 USD
AAJ64242-LB0 CAAAJ64242-LB0
MAC 1753
MAC 1753

A non-peptide GnRH agent capable of inhibiting the effect of gonadotropin-releasing hormone

Formula: C₁₆H₁₀Cl₂N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD02678097
CAS Number: 685830-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR