Lck inhibitor II, solid
Supplier: Thermo Scientific Chemicals
J64111-LB0
J64111.LB0
AAJ64111-LB0EA
323.62
USD
AAJ64111-LB0
CAAAJ64111-LB0
Lck inhibitor II, solid
Lck inhibitor II
Pale yellow Solid
- Light Sensitive
A cell-permeable, ATP binding site-targeting, tri-substituted pyrimidine that acts as a highly potent Lck (lymphocyte specific kinase) inhibitor
Formula:
C₂₄H₂₆N₆O₃ MW: 446.50 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22581248 CAS Number: 918870-43-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...