Antipain dihydrochloride
Supplier: Thermo Scientific
J63680-LB0 J63680-LB0
AAJ63680-LB0EA 124.84 USD
AAJ63680-LB0 CAAAJ63680-LB0
Antipain dihydrochloride
Proteins and Peptides
A natural protease inhibitor for trypsin, papain and cathepsins A and B.
  • Solubility: 0.01 M in water