Thrombin, bovine plasma

Supplier: Alfa Aesar
J63383-EXT J63383-EXP J63383-EXP
AAJ63383-EXTEA 2550.59 USD InStock
AAJ63383-EXT AAJ63383-EXP CAAAJ63383-EXP
Thrombin, bovine plasma
Proteins and Peptides
CAS No.: 9002-04-4
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Reconstitute in 0.9% sodium chloride
Merck Reference: 14,9383
MDL No.: MFCD00166053
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR