Ebselen, powder

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

J63190-MA
10814-278EA 111.82 USD
10814-278
Ebselen, powder
Ebselen

Seleno-organic compound that inhibits nitric oxide-induced apoptosis. Potent antioxidant. Protects calcium influx blockage

Formula: C₁₃H₉NOSe
MW: 274.20 g/mol
Melting Pt: 180…181 °C
Storage Temperature: Freezer
MDL Number: MFCD00210937
CAS Number: 60940-34-3
UN: 3283
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR