Ginkgolide A, Powder
Supplier: Thermo Scientific Chemicals
J62384-FPL
J62384.FPL
AAJ62384-FPLEA
150.14
USD
CAAAJ62384-FPL
AAJ62384-FPL
Ginkgolide A, Powder
Ginkgolide A
Platelet-Activating Factor (PAF) antagonist
Formula:
C₂₀H₂₄O₉ Storage Temperature: Freezer |
MDL Number:
MFCD00133365 CAS Number: 15291-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...