K-252a, Powder

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl-6-hydroxy-5-methyl-13-oxo-5,6,7,8,14,15-hexahydro-13h-5,8-epoxy-4b,8a,14-triazadibenzo[b,h]cycloocta[1,2,3,4-jkl]cyclopenta[e]-as-indacene-6-carboxylate

J61687-LB0 J61687-MCR J61687.MCR
AAJ61687-MCREA 310.45 USD
CAAAJ61687-MCR AAJ61687-MCR CAAAJ61687-LB0
K-252a, Powder
K-252a

Inhibitor of protein kinase A


  • Off-white powder

Formula: C₂₇H₂₁N₃O₅
MW: 467.481 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00161522
CAS Number: 97161-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR