Paxilline ≥97%

Supplier: Thermo Scientific Chemicals
Danger

J61341-LB0 J61341-MB J61341-MB
AAJ61341-LB0EA 263.62 USD
AAJ61341-LB0 CAAAJ61341-MB AAJ61341-MB
Paxilline ≥97%
Paxilline

Potent blocker of high-conductance calcium-activated potassium (BKCa) channels

Formula: C₂₇H₃₃NO₄
Storage Temperature: Freezer
MDL Number: MFCD00083464
CAS Number: 57186-25-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR