Okadaic acid 98%, Powder

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: OA9,10-deepithio-9,10-didehydroacanthifolicin

J60155-EXS
AAJ60155-EXSEA 662.4 USD
AAJ60155-EXS
Okadaic acid 98%, Powder
Okadaic acid

Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A). Does not inhibit tyrosine or alkaline phosphatases

Formula: C₄₄H₆₈O₁₃
MW: 805.02 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00083455
CAS Number: 78111-17-8
UN: 3462
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR