RF40 Battery Coolant/Glycol Refractometer with Automatic Temperature Compensation, Extech®
Supplier: Extech
RF40
89370-230EA 211.71 USD
89370-230
RF40 Battery Coolant/Glycol Refractometer with Automatic Temperature Compensation, Extech®
Refractometers Handheld Refractometers
Portable Battery Coolant/Glycol Refractometer with ATC that measures the freezing point of coolants and concentrations of Ethylene/Propylene Glycol
The unit features Automatic Temperature Compensation (ATC). Requires only 2 or 3 drops of solution. Compact size, easy to operate.
Certifications: CE marked.
Ordering information: Supplied complete with case, calibration screwdriver, and calibration solution. Refractometer also includes manufacturer's one-year warranty.