SENSION+ MM150 DL PORTABLE METER
Supplier: Hach
Multi-Meter^ Sension+ MM150 Portable pH/ORP/EC Data Logger Multi-Meter, 3AA, 0 to 50 degree C, -2 to 19.99 pH, Memory for 500 Measurement points with time and date