The Powerpette Pro pipette controller fits the hand comfortably with the pipette holder at an angle, ideal for use within a laminar flow or microbiological safety cabinet.
- Comfortable, ergonomic design for extended use
- Equipped with powerful and quiet motor
- Multiple operation modes enable a wide variety of liquid handling applications
The Powerpette mode-selection switch enables selection of high or low modes, both allowing variable aspirate and dispense speeds (with blow out). The gravity dispense mode is designed for use with 'to deliver' (TD) pipettes. The speed of suction and dispensing is controlled through the concave finger triggers, designed to provide a comfortable and positive grip requiring minimal effort. Suitable for all types of pipettes 1 to 100 ml. Fills a 25 ml pipette in under three seconds on the fastest setting. The pipette filler can be recharged during use or while housed, with or without the pipette attached. It is supplied with a stand that can be used to hold the Powerpette Pro either on the bench or on the wall.
The controller delivers an efficient performance with a powerful yet quiet motor to speed up large volume pipetting.
Certifications: CE marked.
Ordering information: Unit is fitted with a 0.45 µm filter. Also supplied with manufacturer’s three-year warranty, one 0.45 µm and one 0.2 µm hydrophobic filter, charger, bench stand and colored nose cone set.