Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heavy-Duty Light Meter, Extech® Instruments

Supplier: Extech
HD400-NIST HD400
Heavy-duty light meter features double molded housing and a large backlit display with 40-segment bar graph

The HD400 is supplied complete with a light sensor with 1m (3') cable and protective cover, Windows® compatible software with USB cable, hard carrying case, and 9V battery.

The HD400 utilizes a precision silicon photo diode and spectral response filter. The device also features a peak mode (10mS) that captures the highest reading and a relative mode that indicates change in light levels. The heavy-duty, rugged double molded housing enables use in any environment.

Certifications: CE marked.

Ordering information: Supplied with manufacturer's three-year warranty.



 

ORDER

Expand All / Collapse All
Description Model Supplier No. VWR Catalog Number Unit Price Quantity
Heavy-Duty Light Meter HD400 HD400 89200-748 Each Retrieving
Heavy-Duty Light Meter with certificate of calibration traceable to NIST HD400 HD400-NIST 10171-766 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Resolution 0.01 Fc/0.1 Lux
Basic Accuracy ±5% rdg
Fc Range 40, 400, 4000, 40.00 kFc
Lux Range 400, 4000, 40k, 400 kLux

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty