    Sort by:
  • Search Within Results
1,2-Diamino-9,10-anthraquinone
SearchPresentationType-HORIZONTAL
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (89138-176)
Supplier:  Biotium
Description:   reacts with nitric oxide (NO) to form a red (em > 580 nm) fluorescent precipitate. DAA probe has been used to detect NO concentration in rat retinas after injury to the optic nerve.