    Sort by:
  • Search Within Results
4-tert-Butylphthalic anhydride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 32703-79-0 MDL Number: MFCD00060127 Molecular Formula: C12H12O3 Molecular Weight: 204.23 Purity/Analysis Method: 98.0% (GC,T) For...