You Searched For: 2-Methoxyethyl chloroformate
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: TCI America
Description: CAS Number: 628-12-6 MDL Number: MFCD00058932 Molecular Formula: C4H7ClO3 Molecular Weight: 138.55 Purity/Analysis Method: 93.0% (GC,T) Form:...