    Sort by:
  • Search Within Results
Dinonyl phthalate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Supplier:  Acros Organics
Description:   Dinonyl phthalate, purity: 97%, CAS Number: 84-76-4, Molecular Formula: C26H42O4, Molecular Wight: 418.61, Form: Viscous liquid, color: Clear colorless to yellow-brown, size: 100G