You Searched For: Fast Blue B salt zinc double salt
1  results were found
SearchResultCount:"1"
Sort Results
List View Easy View
Rate These Search Results
Supplier: MP Biomedicals
Description: Fast blue B salt is useful for TLC of Aflatoxins.