Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "samarium-diiodide-in-thf"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Samarium(II) iodide, 0.07 - 0.12M in THF, stab, Cas Number: 32248-43-4, Molecular Formula: SmI2, Color: Dark, Form: Liquid, Size: 500ML