    Sort by:
1,4-Diamino-2,3-dicyano-9,10-anthraquinone
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 81-41-4 MDL Number: MFCD00053082 Molecular Formula: C16H8N4O2 Molecular Weight: 288.27 Purity/Analysis Method: 90.0% (HPLC,N) For...