BAG BIOPSY WHITE SM 1000CS
Supplier: Thermo Scientific
Biopsy Bag^ Biopsy Bag, help reduce the risks of small specimen loss, reagent carryover and pressure artifact. Dimensions: 1.2x1.7 (31x43mm), white, small.