You Searched For: Voltmeters
Voltmeters present a convenient and easy-to-use tool for measuring the voltage. The meters can also be used to calculate the potential in direct-current or alternating-current circuits, and voltages developed in electrochemical or voltaic cells in chemistry applications. Models may offer audible or visual indication of voltage readings. The sensitivity ranges can be adjusted to sense particular voltage levels. All voltmeters are portable and are constructed from durable materials.
Voltmeters present a convenient and easy-to-use tool for measuring the voltage. The meters can also be used to calculate the potential in direct-current or alternating-current circuits, and voltages developed in electrochemical or voltaic cells in chemistry applications. Models may offer audible or visual indication of voltage readings. The sensitivity ranges can be adjusted to sense particular voltage levels. All voltmeters are portable and are constructed from durable materials.
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View
Rate These Search Results
Supplier: VWR International
Description: Full-sized and fully enclosed, these meters are economically priced and offer ±2.5% accuracy.
Catalog Number: (470137-638)
Supplier: VWR International
Description: This versatile galvanometer demonstrates how a current loop generates a magnetic field.
Limited time offers
Supplier: VWR International
Description: Demonstration Meter
Catalog Number: (470006-690)
Supplier: VWR International
Description: Simple and efficient.
Supplier: Transforming Technologies
Description: A convenient hand-held device designed to locate and measure electrostatic fields on surfaces.
Supplier: United Scientific Supplies
Description: Measure the potential difference in DC circuits.
Catalog Number: (470017-076)
Supplier: Vernier Software & Technology
Description: The Vernier Voltage Probe is a convenient and easy way to measure voltage.
Catalog Number: (76474-634)
Supplier: Transforming Technologies
Description: A complete system to measure the body voltage generated while walking.
Catalog Number: (66170-284)
Supplier: LaMotte
Description: This kit tests for Low Iron in water using Iron Reagents and the LRC Comparator
SDS Small Business Enterprise
Catalog Number: (470112-966)
Supplier: Vernier Software & Technology
Description: This sensor is used to measure voltages in the range of -30 to +30 volts.
Catalog Number: (76251-628)
Supplier: DIGI-KEY CORPORATION
Description: Precision Potentiometer with plastic or metal shaft and bushings.
Catalog Number: (470336-122)
Supplier: Wards
Description: Light and portable digital voltmeter used to measure the potential difference between two points.
Catalog Number: (470126-654)
Supplier: United Scientific Supplies
Description: Start your electricity studies with a wide variety of electrical components.
Catalog Number: (10748-278)
Supplier: Avantor
Description: The FLIR CM78 is a True RMS Industrial Grade 1000A Clamp Meter for the electrician who needs a safe, capable combination tool engineered with advanced...
UL Listed
Catalog Number: (14223-826)
Supplier: Statico
Description: The S2001 is designed to locate and measure static charge potentials on product, people, equipment and packaging
Catalog Number: (470110-600)
Supplier: Wards
Description: Rugged Design perfect for student labs.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 16 of 19
no targeter for Bottom