Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Straight Wall Zirconium Crucibles, Kitco Metals Inc.

Supplier: XRF SCIENTIFIC AMERICAS, INC.
ZSW30 ZSW250 ZSW40 ZSW50 ZSW45 ZSW55 ZSW10 ZSW75 ZSW20 ZSW80 ZSW15 ZSW25 ZSW35 ZSW100
Zirconium's distinctive properties make it a popular choice for fusions and fluxes in analytical chemistry and chemical processing.

These straight wall Zirconium crucibles are highly resistant to corrosion and do not require handling with platinum tipped tongs. They can be cleaned with most common agents (except hydrofluoric acid) and suffer little to no attack or oxidation. Zirconium crucibles are resistant to melts of alkali carbonates, hydroxides, peroxides, borates, nitrate chlorides, and some fluorides. This construction virtually eliminates the problem of sample contamination. Lids are available upon request.

Ordering information: Lids are sold separately.



 

ORDER

Expand All / Collapse All
Description Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Zirconium Crucible, Straight Wall 10 mL ZSW10 89401-388 Each Retrieving
Zirconium Crucible, Straight Wall 15 mL ZSW15 89401-390 Each Retrieving
Zirconium Crucible, Straight Wall 20 mL ZSW20 89401-392 Each Retrieving
Zirconium Crucible, Straight Wall 25 mL ZSW25 89401-394 Each Retrieving
Zirconium Crucible, Straight Wall 30 mL ZSW30 89401-396 Each Retrieving
Zirconium Crucible, Straight Wall 35 mL ZSW35 89401-398 Each Retrieving
Zirconium Crucible, Straight Wall 40 mL ZSW40 89401-400 Each Retrieving
Zirconium Crucible, Straight Wall 45 mL ZSW45 89401-402 Each Retrieving
Zirconium Crucible, Straight Wall 50 mL ZSW50 89401-404 Each Retrieving
Zirconium Crucible, Straight Wall 55 mL ZSW55 89401-406 Each Retrieving
Zirconium Crucible, Straight Wall 75 mL ZSW75 89401-408 Each Retrieving
Zirconium Crucible, Straight Wall 80 mL ZSW80 89401-410 Each Retrieving
Zirconium Crucible, Straight Wall 100 mL ZSW100 89401-412 Each Retrieving
Zirconium Crucible, Straight Wall 250 mL ZSW250 89401-414 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty