UREA NITROGEN LIQUI-UV (RATE) 30 ML
Supplier: Stanbio Laboratory
Urea Nitrogen (BUN) Liqui-UV* Test^ Urea Nitrogen (BUN) Liqui-UV* Test (Rate), Modified Urease Methodology, Ready-to-use liquid reagents, R1: 4 x 25 mL, R2: 4 x 5 mL